Merck
  • Epigenetic priming of non-small cell lung cancer cell lines to the antiproliferative and differentiating effects of all-trans retinoic acid.
Epigenetic priming of non-small cell lung cancer cell lines to the antiproliferative and differentiating effects of all-trans retinoic acid.
Journal of cancer research and clinical oncology (2015-05-27)
Gabriele Greve, Insa Schiffmann, Michael Lübbert
ABSTRACT
The retinoic acid signaling pathway, crucial for differentiation, is silenced by epigenetic mechanisms in many cancers. Epigenetically active, chromatin-modifying agents offer a novel treatment approach, by reactivating aberrantly silenced genes in tumor cells and by sensitizing them to subsequent treatments. We hypothesized that the treatment of non-small cell lung cancer (NSCLC) cells with a histone deacetylase (HDAC) inhibitor may prime them to the antiproliferative and differentiating activity of all-trans retinoic acid. The NSCLC cell lines A549, NCI-H460 and HCC827 were treated with ATRA (2 µM) and the pan-HDAC inhibitor panobinostat (LBH589; 10-35 nM). While treatment with ATRA alone showed only very modest effects, panobinostat reduced cellular proliferation by at least 50 %. Notably, the combination of panobinostat and ATRA had additive and synergistic effects, respectively, on growth inhibition and differentiation, with almost no cytotoxicity. Effects were strongest in A549, followed by the EGFR-mutant HCC827, and least pronounced in NCI-H460. Global histone H3 acetylation was strongly induced by panobinostat; interestingly, ATRA alone had also an effect on histone acetylation, which was synergistically enhanced when the HDAC inhibitor was added. The combination of the two drugs additively decreased expression of phospho-ERK and phospho-AKT, whereas p53 and p21(CIP1/WAF1) proteins were both induced. Panobinostat sensitized, to varying degrees, all three cell lines to the antiproliferative and differentiating effects of ATRA, with synergistic histone H3 acetylation. Combination therapy with an epigenetic drug and ATRA may offer an alternative to aggressive chemotherapy even in primary ATRA-insensitive tumors, such as adenocarcinomas of the lung.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture