Merck
  • Whole-Body Cryostimulation Limits Overreaching in Elite Synchronized Swimmers.
Whole-Body Cryostimulation Limits Overreaching in Elite Synchronized Swimmers.
Medicine and science in sports and exercise (2014-10-15)
Karine Schaal, Yann LE Meur, Julien Louis, Jean-Robert Filliard, Philippe Hellard, Gretchen Casazza, Christophe Hausswirth
ABSTRACT
Elite athletes frequently undergo periods of intensified training (IT) within their normal training program. These periods can lead athletes into functional overreaching, characterized by high perceived fatigue, impaired sleep, and performance. Because whole-body cryostimulation (WBC) has been proven to be an effective recovery method in the short term (<76 h), we investigated whether daily WBC sessions during IT could prevent exercise and sleep-related signs of overreaching. After a normal training week (BASE), 10 elite synchronized swimmers performed two 2-wk IT periods in a randomized crossover fashion using WBC daily (ITWBC) or not (ITCON), separated by 9 d of light training. Swim time trials (400 m) were performed at BASE and after each IT to quantify blood lactate ([La]B), HR (HR400), salivary alpha amylase ([α-amylase]s400), and cortisol ([cortisol]s400) responses. Swimmers wore a wrist actigraph nightly to monitor sleep patterns. Swim speed (400 m), [La]B400, and [α-amylase]s400 decreased from BASE to ITCON, although no significant changes were found after ITWBC. Decreased swim speed was correlated to decreased HR400 and [cortisol]s400. During ITCON, significant decreases in actual sleep duration (-21 ± 7 min) and sleep efficiency (-1.9% ± 0.8%) were observed, with increased sleep latency (+11 ± 5 min) and fatigue compared with BASE, although these variables did not change during ITWBC. Using a qualitative statistical analysis, we observed that daily WBC use resulted in a 98%, 59%, 66%, and 78% chance of preserving these respective variables compared with ITCON. WBC use during IT helped mitigate the signs of functional overreaching observed during ITCON, such as reduced sleep quantity, increased fatigue, and impaired exercise capacity. These results support the daily use of WBC by athletes seeking to avoid functional overreaching during key periods of competition preparation.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)