Merck
  • Determination of high-intensity sweeteners in river water and wastewater by solid-phase extraction and liquid chromatography-tandem mass spectrometry.
Determination of high-intensity sweeteners in river water and wastewater by solid-phase extraction and liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2015-04-05)
Paula Arbeláez, Francesc Borrull, Eva Pocurull, Rosa Maria Marcé
ABSTRACT
High-intensity sweeteners have been suggested as potential organic contaminants due to their widespread use in food, drugs and sanitary products. As a consequence, they are introduced into the environment by different pathways, affecting aquatic life. In this study, a method based on solid-phase extraction (SPE) and liquid chromatography-tandem mass spectrometry (LC-MS-MS) has been developed and validated for the determination of eight sweeteners (saccharin, cyclamate, aspartame, acesulfame, neohesperidin dihydrochalcone, sucralose, stevioside and glycyrrhizic acid) in river water and wastewater. To get the maximum recoveries in SPE, several commercial sorbents were tested and Oasis HLB gave the best results, with recoveries higher than 41% for all of the compounds in the different matrices. Method limits of detection were in the range of 0.001-0.04μg/L in river water and 0.01-0.5μg/L in influent and effluent wastewater. Method reproducibility between days (n=5) was below 15% for all compounds. The method was applied to the determination of sweeteners in various river waters and wastewaters in Catalonia. Cyclamate, aspartame, neohesperidin dihydrochalcone, acesulfame and sucralose were found in river water, with the two last compounds being present at the highest values (1.62μg/L for acesulfame and 3.57μg/L for sucralose). In influent and effluent wastewater, all of the compounds were found at concentration levels ranging from 0.05 to 155μg/L except for stevioside and neohesperidin dihydrochalcone, which were not detected.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥95.0% (NT)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥70% (HPLC)
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%