Merck
  • A novel ultrasound-assisted back extraction reverse micelles method coupled with gas chromatography-flame ionization detection for determination of aldehydes in heated edibles oils.
A novel ultrasound-assisted back extraction reverse micelles method coupled with gas chromatography-flame ionization detection for determination of aldehydes in heated edibles oils.
Food chemistry (2015-06-05)
Zahra Ramezani, Roya Mirzajani, Fatemeh Kardani
ABSTRACT
A novel ultrasound-assisted back extraction reverse micelles coupled with gas chromatography-flame ionization detection has been developed for the extraction and determination of some short chain aldehydes in different heated edible oil samples. After the homogenization of the oil samples with Triton X-100, 200 μL of methanol was added to facilitate the phase separation. The aqueous micelle phase has been separated by centrifugation, then it was treated with a mixture of H2O: CHCl3 and ultrasonic vibration, were used to effectively back-extraction of the analytes into the chloroform phase. The sedimented organic phase was obtained after centrifugation, withdrawn into the microsyringe and directly injected into the GC-FID system. The calibration graphs were linear in the range 0.05-20 mg L(-1). The limits of detection were in the range of 0.02-0.15 mg L(-1). This procedure was successfully applied for determination of propanal, butanal, hexanal and heptanal in real heated oil samples.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Butyraldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Heptaldehyde, 95%
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Sigma-Aldrich
Butyraldehyde, ≥96.0%
Sigma-Aldrich
Butyraldehyde, natural, FG
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Hexanal, ≥97%, FCC, stabilized, FG
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O