Merck
  • Wheat Type (Class) Influences Development and Regression of Colon Cancer Risk Markers in Rats.
Wheat Type (Class) Influences Development and Regression of Colon Cancer Risk Markers in Rats.
Nutrition and cancer (2015-10-17)
Ajmila Islam, Daniel D Gallaher
ABSTRACT
We previously found red wheat more effective than white wheat in reducing colon cancer risk in rats when fed during initiation and postinitiation stages. Here we examine the effect of wheat on colon cancer risk in early and late postinitiation stages in carcinogen-treated rats. Four groups were fed a basal diet, 1 group a red wheat diet, and 1 group a white wheat diet. After 6 wk, 1 basal, the red and white groups were killed (early postinitiation stage). Of the remaining basal groups, 1 continued on the basal diet, 1 was switched to red and another to white wheat for 8 more wk (late postinitiation stage). Red and white wheat significantly reduced morphological [aberrant crypt foci (ACF)] and biochemical (β-catenin accumulated crypts) markers in both early and late postinitiation stages. Both wheat diets reduced dysplasia markers (sialomucin-expressing ACF and mucin depleted foci), compared to the basal diet, during the late postinitiation stage, but red wheat more so. Only red wheat significantly reduced the number of metallothionein-positive crypts, a stem cell mutation marker, in both stages. Overall, red wheat flour reduced risk markers more than white wheat flour, and this was more pronounced in the late post-initiation stage.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Pricing and availability is not currently available.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Pricing and availability is not currently available.
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Pricing and availability is not currently available.
SAFC
L-Cystine
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Cystine, Wacker Chemie AG, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Choline bitartrate
Pricing and availability is not currently available.
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Pricing and availability is not currently available.
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Pricing and availability is not currently available.