Merck
  • Topical delivery of dexamethasone acetate from hydrogel containing nanostructured liquid carriers and the drug.
Topical delivery of dexamethasone acetate from hydrogel containing nanostructured liquid carriers and the drug.
Archives of pharmacal research (2015-05-06)
Nguyen-Thach Tung, Vu-Thu Huyen, Sang-Cheol Chi
ABSTRACT
The potential of hydrogel containing nanostructured lipid carriers (NLC) to enhance the skin permeation rate and skin deposition of dexamethasone acetate (DEA) was investigated. The particle size of obtained NLCs was around 224.4 nm. NLCs had core-shell structure and DEA existed in amorphous state in NLCs. The permeation rate of DEA through excised mouse skins from hydrogel containing DEA-NLC (DEA-NLC-hydrogel) was 7.3 times higher than DEA-ointment. The skin deposition of DEA from DEA-NLC-hydrogel increased 3.8 folds compared to that from solution of DEA in hydrogel (DEA-hydrogel).
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Triton X-100, laboratory grade