Merck
  • Ricinus communis L. stem bark extracts regulate ovarian cell functions and secretory activity and their response to Luteinising hormone.
Ricinus communis L. stem bark extracts regulate ovarian cell functions and secretory activity and their response to Luteinising hormone.
International journal of impotence research (2015-08-28)
S Nath, A Kadasi, R Grossmann, A V Sirotkin, A Kolesarova, A D Talukdar, M D Choudhury
ABSTRACT
Ricinus communis L. has ethnopharmacological contraceptive reputation but its stem bark has unexplored mechanisms of action in female reproductive system. In the present study, the effect of methanolic and aqueous extracts from the stem bark of the plant was examined on basic porcine ovarian granulosa cell functions and its response to Luteinising hormone (LH)-the upstream hormonal regulator. Systemic treatment of methanolic and aqueous extracts stimulated cell proliferation (proliferating cell nuclear antigen, PCNA) and also promoted cell apoptosis (caspase-3). Aqueous extract has inverted the stimulatory effect of LH on PCNA but not on caspase-3. Methanolic extract stimulated as well as inhibited progesterone release and stimulated testosterone secretion. Whereas aqueous extract inhibited both steroid releases and suppressed the stimulatory effect of LH on progesterone release and promoted the inhibitory effect of LH on testosterone release. In conclusion, the present study unveils the mechanism of action of R. communis stem bark in in vitro condition. These suggest its possible contraceptive efficacy by exerting its regulatory role over LH and on basic ovarian cell functions and secretion activity.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.