Merck
  • Formation of fluorescent polydopamine dots from hydroxyl radical-induced degradation of polydopamine nanoparticles.
Formation of fluorescent polydopamine dots from hydroxyl radical-induced degradation of polydopamine nanoparticles.
Physical chemistry chemical physics : PCCP (2015-03-31)
Jia-Hui Lin, Cheng-Ju Yu, Ya-Chun Yang, Wei-Lung Tseng
ABSTRACT
This study describes the synthesis of fluorescent polydopamine dots (PDs) through hydroxyl radical-induced degradation of polydopamine nanoparticles. The decomposition of polydopamine nanoparticles to fluorescent PDs was confirmed using transmission electron microscopy and dark-field microscopy. The analysis of PDs by using laser desorption/ionization time-of-flight mass spectrometry revealed that the PDs consisted of dopamine, 5,6-dihydroxyindole, and trihydroxyindole units. Oligomerization and self-assembly of these units produced a broad adsorption band, resulting in an excitation-wavelength-dependent emission behavior. The maximal fluorescence of PDs appeared at 440 nm with a quantum yield of 1.2%. The coordination between the catechol groups of PDs and ferric ions (Fe(3+)) quenched the fluorescence of PDs; the limit of detection at a signal-to-noise ratio of 3 for Fe(3+) was determined to be 0.3 μM. The presence of pyrophosphate switched on the fluorescence of the PD-Fe(3+) complexes. Compared to the other reported methods for sensing Fe(3+), PDs provided simple, low-cost, and reusable detection of Fe(3+).
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Nickel(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel(II) chloride, 98%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
SAFC
Tromethamine
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Trizma® base, ≥99.0% (T)