Merck
  • Combinatorial BTK and MALT1 inhibition augments killing of CD79 mutant diffuse large B cell lymphoma.
Combinatorial BTK and MALT1 inhibition augments killing of CD79 mutant diffuse large B cell lymphoma.
Oncotarget (2015-11-06)
Daniel Nagel, Miriam Bognar, Andrea C Eitelhuber, Kerstin Kutzner, Michelle Vincendeau, Daniel Krappmann
ABSTRACT
Survival of activated B cell-subtype (ABC) of diffuse large B cell lymphoma (DLBCL) is driven by chronic B cell receptor (BCR) signaling that activates the canonical NF-κB pathway. Inhibition of BTK by Ibrutinib has been shown to kill ABC DLBCL cells that carry activating mutations in the BCR adaptor CD79. However, mutations in BTK or in downstream components such as CARMA1/CARD11 can render lymphomas Ibrutinib resistant. Therefore, we assessed here the simultaneous inhibition of BTK and the protease MALT1 that acts downstream of CARMA1 and is essential for ABC DLBCL tumor growth. We show that in CD79 mutant cells BTK is a crucial upstream regulator of MALT1, but dispensable in CARMA1 mutant ABC DLBCL. Combined inhibition of BTK by Ibrutinib and MALT1 by S-Mepazine additively impaired MALT1 cleavage activity and expression of NF-κB pro-survival factors. Thereby, combinatorial Ibrutinib and S-Mepazine treatment enhanced killing of CD79 mutant ABC DLBCL cells. Moreover, while expression of oncogenic CARMA1 in CD79 mutant cells conferred Ibrutinib resistance, double mutant cells were still sensitive to MALT1 inhibition by S-Mepazine. Thus, based on the genetic background combinatorial BTK and MALT1 inhibition may improve effectiveness of therapeutic treatment and reduce the chances for the development of drug resistances.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)