Merck
  • NF1 loss induces senescence during human melanocyte differentiation in an iPSC-based model.
NF1 loss induces senescence during human melanocyte differentiation in an iPSC-based model.
Pigment cell & melanoma research (2015-04-01)
Lionel Larribere, Huizi Wu, Daniel Novak, Marta Galach, Mathias Bernhardt, Elias Orouji, Kasia Weina, Nathalie Knappe, Christos Sachpekidis, Ludmila Umansky, Philipp Beckhove, Viktor Umansky, Sofie De Schepper, Dieter Kaufmann, Robert Ballotti, Corine Bertolotto, Jochen Utikal
ABSTRACT
Neurofibromatosis type 1 (NF1) is a frequent genetic disease leading to the development of Schwann cell-derived neurofibromas or melanocytic lesions called café-au-lait macules (CALMs). The molecular mechanisms involved in CALMs formation remain largely unknown. In this report, we show for the first time pathophysiological mechanisms of abnormal melanocyte differentiation in a human NF1(+/-) -induced pluripotent stem cell (iPSC)-based model. We demonstrate that NF1 patient-derived fibroblasts can be successfully reprogrammed in NF1(+/-) iPSCs with active RAS signaling and that NF1 loss induces senescence during melanocyte differentiation as well as in patient's-derived CALMs, revealing a new role for NF1 in the melanocyte lineage.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
MISSION® esiRNA, targeting human NF1
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nf1
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %