Merck
  • LC-APCI-MS/MS Quantification and Topical Bioavailability of Chloroacetamide in Rats.
LC-APCI-MS/MS Quantification and Topical Bioavailability of Chloroacetamide in Rats.
Journal of chromatographic science (2014-12-20)
Tae Hwan Kim, Su Hyun Seok, Kyu-Bong Kim, Beom Soo Shin, Jeong Pyo Lee, Yong-Kyu Choi, Min Gi Kim, Min Kyu Kim, Sang Min Kim, Sun Dong Yoo
ABSTRACT
Chloroacetamide (CAA) is a preservative used in various cosmetic, personal care and household products. Due to the hazard potential for allergic reaction and reproductive toxicity, CAA is being considered a high priority for screening assessment and toxicological re-evaluation. This study describes the development of a highly specific and sensitive atmospheric pressure chemical ionization tandem mass spectrometry method for the determination of CAA in rat plasma and its application to a topical bioavailability study. Chromatographic separations were achieved on a C8 column using a highly aqueous mobile phase with a binary gradient elution. The assay was linear in the concentration range of 5-2,500 ng/mL (r ≥ 0.995) using a small sample volume (100 µL). Applicability of the assay was demonstrated in a bioavailability study in rats after i.v. injection (0.5 or 2 mg/kg) and topical application (7.02 mg/kg). Average elimination half-life and clearance ranged from 26.6 to 30.5 min and 53.9 to 57.3 mL/min/kg, respectively. Upon topical application, CAA was slowly but steadily absorbed for a prolonged time period (12 h). The topical bioavailability was 53.5 and 48.3% for emulsion and lotion, respectively. The developed assay may be useful to examine the relationship between exposure and toxic potential of CAA in risk assessment.
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
1-Stearoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
D-Sorbitol, 99% (GC)