Merck
Boosting riboswitch efficiency by RNA amplification.
Nucleic acids research (2015-04-01)
Masoumeh Emadpour, Daniel Karcher, Ralph Bock
ABSTRACT
Riboswitches are RNA sensors that regulate gene expression in response to binding of small molecules. Although they conceptually represent simple on/off switches and, therefore, hold great promise for biotechnology and future synthetic biology applications, the induction of gene expression by natural riboswitches after ligand addition or removal is often only moderate and, consequently, the achievable expression levels are not very high. Here, we have designed an RNA amplification-based system that strongly improves the efficiency of riboswitches. We have successfully implemented the method in a biological system for which currently no efficient endogenous tools for inducible (trans)gene expression are available: the chloroplasts of higher plants. We further show that an HIV antigen whose constitutive expression from the chloroplast genome is deleterious to the plant can be inducibly expressed under the control of the RNA amplification-enhanced riboswitch (RAmpER) without causing a mutant phenotype, demonstrating the potential of the method for the production of proteins and metabolites that are toxic to the host cell.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O