Merck
  • A novel in vitro assay for assessing efficacy and toxicity of antifungals using human leukaemic cells infected with Candida albicans.
A novel in vitro assay for assessing efficacy and toxicity of antifungals using human leukaemic cells infected with Candida albicans.
Journal of applied microbiology (2015-04-08)
J Boros-Majewska, Ł Turczyk, X Wei, S Milewski, D W Williams
ABSTRACT
This study describes a novel in vitro assay that simultaneously determines antifungal efficiency and host cell toxicity using suspensions of human leukaemic cells (HL-60) infected with Candida albicans. The effect of Candida infection on host cell viability was evaluated by the microscopy of trypan blue-stained cells and lactate dehydrogenase (LDH) activity. The in vitro 'drug potency assay' utilized the Cell Counting Kit-8 and measured post-antifungal treatment viability of Candida-infected HL-60 cells and the ability of the antifungal treatment to prevent infection. LDH activity showed that 42% ± 4·0 and 85·3% ± 7·40 of HL-60 cells were killed following Candida infection at the multiplicity of infection (MOI) of 1 : 1 and 1 : 5, respectively. The antifungal nystatin (0·78-25 μmol l(-1) ) was found to inhibit C. albicans infection as seen by the significantly increased viability of HL-60 cells. Cytotoxicity of nystatin towards infected HL-60 cells was evident at higher concentrations and this was also confirmed by propidium iodide staining. An assay using undisturbed cell suspension conditions was successfully developed for assessing the selectivity of the antifungal therapy in the host-Candida environment. The assay employing Candida infection of host cell suspensions represents a promising method for testing interactions of antifungal compounds with both fungal and host cells.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%