Merck
  • A validated method for simultaneous determination of codeine, codeine-6-glucuronide, norcodeine, morphine, morphine-3-glucuronide and morphine-6-glucuronide in post-mortem blood, vitreous fluid, muscle, fat and brain tissue by LC-MS.
A validated method for simultaneous determination of codeine, codeine-6-glucuronide, norcodeine, morphine, morphine-3-glucuronide and morphine-6-glucuronide in post-mortem blood, vitreous fluid, muscle, fat and brain tissue by LC-MS.
Journal of analytical toxicology (2015-01-06)
Joachim Frost, Trine N Løkken, Wenche R Brede, Solfrid Hegstad, Ivar S Nordrum, Lars Slørdal
ABSTRACT
The toxicodynamics and, to a lesser degree, toxicokinetics of the widely used opiate codeine remain a matter of controversy. To address this issue, analytical methods capable of providing reliable quantification of codeine metabolites alongside codeine concentrations are required. This article presents a validated method for simultaneous determination of codeine, codeine metabolites codeine-6-glucuronide (C6G), norcodeine and morphine, and morphine metabolites morphine-3-glucuronide (M3G) and morphine-6-glucuronide (M6G) in post-mortem whole blood, vitreous fluid, muscle, fat and brain tissue by high-performance liquid chromatography mass spectrometry. Samples were prepared by solid-phase extraction. The validated ranges were 1.5-300 ng/mL for codeine, norcodeine and morphine, and 23-4,600 ng/mL for C6G, M3G and M6G, with exceptions for norcodeine in muscle (3-300 ng/mL), morphine in muscle, fat and brain (3-300 ng/mL) and M6G in fat (46-4,600 ng/mL). Within-run and between-run accuracy (88.1-114.1%) and precision (CV 0.6-12.7%), matrix effects (CV 0.3-13.5%) and recovery (57.8-94.1%) were validated at two concentration levels; 3 and 150 ng/mL for codeine, norcodeine and morphine, and 46 and 2,300 ng/mL for C6G, M3G and M6G. Freeze-thaw and long-term stability (6 months at -80°C) was assessed, showing no significant changes in analyte concentrations (-12 to +8%). The method was applied in two authentic forensic autopsy cases implicating codeine in both therapeutic and presumably lethal concentration levels.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.