Merck
  • Enantioselective determination of metoprolol and its metabolites in human urine high-performance liquid chromatography with fluorescence detection (HPLC-FLD) and tandem mass spectrometry (MS/MS).
Enantioselective determination of metoprolol and its metabolites in human urine high-performance liquid chromatography with fluorescence detection (HPLC-FLD) and tandem mass spectrometry (MS/MS).
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-10-10)
Irena Baranowska, Weronika Adolf, Sylwia Magiera
ABSTRACT
A sensitive, stereoselective assay using solid phase extraction and high-performance liquid chromatography (HPLC) with fluorescence detection (FLD) was developed and validated for the analysis of enantiomers of metoprolol and its metabolites (α-hydroxymetoprolol, O-desmethylmetoprolol). Chiral separation was achieved using a CHIRALCEL OD-RH column, packed with cellulose tris-(3,5-dimethylphenyl-carbamate) stationary phase, employing a mobile phase composed by a mixture of 0.2% diethylamine in water and acetonitrile in gradient elution mode. Linear calibration curves were obtained over the range of 0.025-2.0μg/mL (R(2)>0.994) in urine for both enantiomers of metoprolol and its metabolites with quantitation limit of 0.025μg/mL. Intra and inter-day precision and accuracy were below 15% for both metoprolol and metabolites enantiomers. The recovery of enantiomer of metoprolol and its metabolite was greater than 68.0%, utilizing a SPE procedure. The method was tested with urine quality control samples and human urine fractions after administration of 50mg rac-metoprolol.
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Diethylamine, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Diethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Diethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis