Merck
  • Production of Toxocara cati TES-120 Recombinant Antigen and Comparison with its T. canis Homolog for Serodiagnosis of Toxocariasis.
Production of Toxocara cati TES-120 Recombinant Antigen and Comparison with its T. canis Homolog for Serodiagnosis of Toxocariasis.
The American journal of tropical medicine and hygiene (2015-06-03)
Farzaneh Zahabiun, Seyed Mahmoud Sadjjadi, Muhammad Hafiznur Yunus, Anizah Rahumatullah, Mohammad Hosein Falaki Moghaddam, Syazwan Saidin, Rahmah Noordin
ABSTRACT
Toxocariasis is a cosmopolitan zoonotic disease caused by the infective larvae of Toxocara canis and T. cati. Diagnosis in humans is usually based on clinical symptoms and serology. Immunoglobulin G (IgG)-enzyme-linked immunosorbent assay kits using T. canis excretory-secretory (TES) larval antigens are commonly used for serodiagnosis. Differences in the antigens of the two Toxocara species may influence the diagnostic sensitivity of the test. In this study, T. cati recombinant TES-120 (rTES-120) was cloned, expressed, and compared with its T. canis homolog in an IgG4-western blot. The diagnostic sensitivity and specificity of T. cati rTES-120 were 70% (33/47) and 100% (39/39), respectively. T. canis rTES-120 showed 57.4% sensitivity and 94.4% specificity. When the results of assays using rTES-120 of both species were considered, the diagnostic sensitivity was 76%. This study shows that using antigens from both Toxocara species may improve the serodiagnosis of toxocariasis.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
SAFC
Glycine
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP11C
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside