Merck
  • Occurrence of pharmaceuticals in municipal wastewater, in the recipient water, and sedimented particles of northern Lake Päijänne.
Occurrence of pharmaceuticals in municipal wastewater, in the recipient water, and sedimented particles of northern Lake Päijänne.
Environmental science and pollution research international (2015-07-04)
Petra C Lindholm-Lehto, Heidi S J Ahkola, Juha S Knuutinen, Sirpa H Herve
ABSTRACT
The presence of five different pharmaceuticals, consisting of four anti-inflammatory and one antiepileptic drug, was determined in influent and effluent of a municipal wastewater treatment plant (WWTP) near the city of Jyväskylä, Finland, and in the receiving water, northern Lake Päijänne. In addition, samples of sedimented particles were collected among water samples from five locations near the discharge point of the treated wastewater. The solid phase extracts (SPEs) of water samples were analyzed by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) in the multiple reaction monitoring mode. The studied pharmaceuticals were detected from influent, effluent, and lake water but also in the sedimented particles. The concentrations of carbamazepine, diclofenac, ibuprofen, ketoprofen, and naproxen in Lake Päijänne ranged from 1 to 21 ng L(-1), 4 to 209 ng L(-1), 5 to 836 ng L(-1), 9 to 952 ng L(-1), and 2 to 129 ng L(-1), respectively. The concentrations of ketoprofen in sedimented particles ranged from 79 to 135 μg g(-1) while only trace amounts of other selected pharmaceuticals were detected. The results indicate that the concentrations of pharmaceuticals are affected by the biological and chemical reactions occurring in the wastewater treatment processes but also by the UV light in the photic layer of Lake Päijänne. It can be concluded that considerable amount of selected pharmaceuticals are present in the influent and effluent of municipal WWTP but also in the water phase and sedimented particles of northern Lake Päijänne.
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ibuprofen
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%