Merck
  • Extraction of trace nitrophenols in environmental water samples using boronate affinity sorbent.
Extraction of trace nitrophenols in environmental water samples using boronate affinity sorbent.
Analytica chimica acta (2015-11-10)
Yong Zhang, Meng Mei, Xiaojia Huang, Dongxing Yuan
ABSTRACT
In this research, the applicability of a new sorbent based on boronate affinity material is demonstrated. For this purpose, six strong polar nitrophenols were selected as models which are difficult to be extracted in neutral form (only based on hydrophobic interactions). The extracted nitrophenols were separated and determined by high-performance liquid chromatography with diode array detection. The sorbent was synthesized by in situ copolymerization of 3-acrylamidophenylboronic acid and divinylbenzene using dimethyl sulfoxide and azobisisobutyronitrile as porogen solvent and initiator, respectively. The effect of the preparation parameters in the polymerization mixture on extraction performance was investigated in detail. The size and morphology of the sorbent have been characterized via different techniques such as infrared spectroscopy, elemental analysis, scanning electron microscopy and mercury intrusion porosimetry. The important parameters influencing the extraction efficiency were studied and optimized thoroughly. Under the optimum extraction conditions, the limits of detection (S/N = 3) and limits of quantification (S/N = 10) for the target nitrophenols were 0.097-0.28 and 0.32-0.92 μg/L, respectively. The precision of the proposed method was evaluated in terms of intra- and inter-assay variability calculated as RSD, and it was found that the RSDs were all below 9%. Finally, the developed method was successfully applied for environmental water samples such as wastewater, tap, lake and river water. The recoveries varied within the range of 71.2-115% with RSD below 11% in all cases. The results well demonstrate that the new boronate affinity sorbent can extract nitrophenols effectively through multi-interactions including boron-nitrogen coordination, hydrogen-bond and hydrophobic interactions between sorbent and analytes.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)