Merck
  • 3-demethoxy-3-glycosylaminothiocolchicines: Synthesis of a new class of putative muscle relaxant compounds.
3-demethoxy-3-glycosylaminothiocolchicines: Synthesis of a new class of putative muscle relaxant compounds.
Journal of medicinal chemistry (2006-09-01)
Maria Luisa Gelmi, Donato Pocar, Guido Pontremoli, Sara Pellegrino, Ezio Bombardelli, Gabriele Fontana, Antonella Riva, Walter Balduini, Silvia Carloni, Mauro Cimino, Francis Johnson
ABSTRACT
A novel class of 3-demethoxy-3-glycosylaminothiocolchicines (7) was prepared and tested for muscle relaxant activity. The syntheses were performed starting from the new 3-amino-3-demethoxythiocolchicine (5) prepared in good yield from 3-O-demethylthiocolchicine (1c) using the Buchwald-Hartwig reaction. The condensation of 5 with a series of pentose and hexose sugars (6) gave a series of 3-demethoxy-3-glycosylaminothiocolchicines (7). Their preparation was accomplished by adapting and improving a previous procedure for the preparation of N-arylglycosylamines. In particular, replacing traditional heating with microwave irradiation represents the key improvement of the process. The biological activity of the 3-demethoxy-3-glycosylaminothiocolchicines (7) was evaluated on GABA and strychnine-sensitive glycine receptors present in rat brain and spinal cord.
Sigma-Aldrich
Glycine 1 M solution
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
rac-BINAP, 97%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%