Merck
  • Modulation of sarcoplasmic reticulum Ca(2+)-ATPase by chronic and acute exposure to peroxynitrite.
Modulation of sarcoplasmic reticulum Ca(2+)-ATPase by chronic and acute exposure to peroxynitrite.
European journal of biochemistry (2004-06-23)
Yolanda Gutiérrez-Martín, Francisco J Martín-Romero, Francisco A Iñesta-Vaquera, Carlos Gutiérrez-Merino, Fernando Henao
ABSTRACT
The Ca(2+)-ATPase of skeletal muscle sarcoplasmic reticulum (SERCA), an integral membrane protein, becomes irreversibly inactivated in vitro by the addition of a single bolus of peroxynitrite with a K(0.5) of 200-300 microm, and this results in a large decrease of the ATP-dependent Ca2+ gradient across the sarcoplasmic reticulum (SR) membranes. The inactivation of SERCA is raised by treatment of SR vesicles with repetitive micromolar pulses of peroxynitrite. The inhibition of the SERCA is due to the oxidation of thiol groups and tyrosine nitration. Scavengers that react directly with peroxynitrite, such as cysteine, reduced glutathione, NADH, methionine, ascorbate or Trolox, a water-soluble analog of alpha-tocopherol, afforded significant protection. However, dimethyl sulfoxide and mannitol, two hydroxyl radical scavengers, and alpha-tocopherol did not protect SERCA from inactivation. Our results showed that the target of peroxynitrite is the cytosolic globular domain of the SERCA and that major skeletal muscle intracellular reductants (ascorbate, NADH and reduced glutathione) protected against inhibition of this ATPase by peroxynitrite.
Sigma-Aldrich
Anti-Nitrotyrosine Mouse mAb (CC22.8C7.3), liquid, clone CC22.8C7.3, Calbiochem®