Merck
  • Targeted and untargeted characterisation of free and glycosylated simple phenols in cocoa beans using high resolution-tandem mass spectrometry (Q-Orbitrap).
Targeted and untargeted characterisation of free and glycosylated simple phenols in cocoa beans using high resolution-tandem mass spectrometry (Q-Orbitrap).
Journal of chromatography. A (2016-12-19)
C Barnaba, T Nardin, A Pierotti, M Malacarne, R Larcher
ABSTRACT
Free simple phenols have positive effects on health and influence the organoleptic profile of cocoa products, contributing towards defining their aroma and nutritional properties. Glycosidically bound simple phenols can be hydrolysed during the production phase to the corresponding free forms, and thus potentially contribute to the final sensory profile. In this work, 60 samples of Forastero cocoa beans from all over the world were analysed, combining on-line solid phase extraction (SPE) clean-up with ultra-high performance liquid chromatography (UHPLC), coupled with high resolution mass spectrometry. Operating in negative ion mode and with a heated electrospray, 62 simple phenols were measured, of which four were glucosidic precursors, with quantification limits ranging from 0.04 to 40mgkg
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard