Merck
  • Exon- and contraction-dependent functions of titin in sarcomere assembly.
Exon- and contraction-dependent functions of titin in sarcomere assembly.
Development (Cambridge, England) (2016-11-12)
Yu-Huan Shih, Alexey V Dvornikov, Ping Zhu, Xiao Ma, Maengjo Kim, Yonghe Ding, Xiaolei Xu
ABSTRACT
Titin-truncating variants (TTNtvs) are the major cause of dilated cardiomyopathy (DCM); however, allelic heterogeneity (TTNtvs in different exons) results in variable phenotypes, and remains a major hurdle for disease diagnosis and therapy. Here, we generated a panel of ttn mutants in zebrafish. Four single deletion mutants in ttn.2 or ttn.1 resulted in four phenotypes and three double ttn.2/ttn.1 mutants exhibited more severe phenotypes in somites. Protein analysis identified ttn
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid