Merck
  • Histone deacetylase inhibitors suppress TF-kappaB-dependent agonist-driven tissue factor expression in endothelial cells and monocytes.
Histone deacetylase inhibitors suppress TF-kappaB-dependent agonist-driven tissue factor expression in endothelial cells and monocytes.
The Journal of biological chemistry (2007-08-07)
Jianguo Wang, Shawn A Mahmud, Peter B Bitterman, Yuqing Huo, Arne Slungaard
ABSTRACT
Histone deacetylase inhibitors (HDACi), such as trichostatin A (TSA), can regulate gene expression by promoting acetylation of histones and transcription factors. Human tissue factor (TF) expression is partly governed by a unique, NF-kappaB-related "TF-kappaB" promoter binding site. We find that TSA and four other HDACi (apicidin, MS-275, sodium butyrate, and valproic acid) all inhibit by approximately 90% TF activity and protein level induction in human umbilical vein endothelial cells stimulated by the physiologic agonists tumor necrosis factor (TNF)-alpha, interleukin-1beta, lipopolysaccharide, and HOSCN without affecting expression of the NF-kappaB-regulated adhesion molecules ICAM-1 and E-selectin. TSA and butyrate also blunt TF induction approximately 50% in vitro in peripheral blood mononuclear cells and in vivo in thioglycolate-elicited murine peritoneal macrophages. In human umbilical vein endothelial cells, TSA attenuates by approximately 70% TNF-alpha stimulation of TF mRNA transcription without affecting that of ICAM-1. By electrophoretic mobility shift assay analyses, TNF-alpha and lipopolysaccharide induce strong p65/p50 and p65/c-Rel heterodimer binding to both NF-kappaB and TF-kappaB probes. TSA nearly abolishes TF-kappaB binding without affecting NF-kappaB binding. A chromatin immunoprecipitation assay and a promoter-luciferase reporter system confirm that TSA inhibits TF-kappaB but not NF-kappaB activation. Chromatin immunoprecipitation and small interfering RNA inhibitor studies demonstrate that HDAC3 plays a significant role in TNF-alpha-mediated TF induction. Thus, HDACi transcriptionally inhibit agonist-induced TF expression in endothelial cells and monocytes by a TF-kappaB- and HDAC3-dependent mechanism. We conclude that histone deacetylases, particularly HDAC3, play a hitherto unsuspected role in regulating TF expression and raise the possibility that HDACi might be a novel therapy for thrombotic disorders.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.