Skip to Content
AR
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Ca
Ca
EMBO reports (2017-05-17)
Valentina Giorgio, Victoria Burchell, Marco Schiavone, Claudio Bassot, Giovanni Minervini, Valeria Petronilli, Francesco Argenton, Michael Forte, Silvio Tosatto, Giovanna Lippe, Paolo Bernardi
PMID
28507163
ABSTRACT
F-ATP synthases convert the electrochemical energy of the H
Product Description
G5500
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Pricing
X100
Sigma-Aldrich
Triton
™
X-100, laboratory grade
Pricing
EHU029131
Sigma-Aldrich
MISSION
®
esiRNA, targeting human ATP5B
Pricing