Merck
  • Simultaneous determination of chromones and coumarins in Radix Saposhnikoviae by high performance liquid chromatography with diode array and tandem mass detectors.
Simultaneous determination of chromones and coumarins in Radix Saposhnikoviae by high performance liquid chromatography with diode array and tandem mass detectors.
Journal of chromatography. A (2011-08-03)
Min Kyung Kim, Dong-Hyug Yang, Mihye Jung, Eun Ha Jung, Han Young Eom, Joon Hyuk Suh, Jung Won Min, Unyong Kim, Hyeyoung Min, Jinwoong Kim, Sang Beom Han
ABSTRACT
Methods using high performance liquid chromatography with diode array detection (HPLC-DAD) and tandem mass spectrometry (HPLC-MS/MS) were developed and validated for the simultaneous determination of 5 chromones and 6 coumarins: prim-O-glucosylcimifugin (1), cimifugin (2), nodakenin (3), 4'-O-β-d-glucosyl-5-O-methylvisamminol (4), sec-O-glucosylhamaudol (5), psoralen (6), bergapten (7), imperatorin (8), phellopterin (9), 3'-O-angeloylhamaudol (10) and anomalin (11), in Radix Saposhnikoviae. The separation conditions for HPLC-DAD were optimized using an Ascentis Express C18 (4.6 mm×100 mm, 2.7 μm particle size) fused-core column. The mobile phase was composed of 10% aqueous acetonitrile (A) and 90% acetonitrile (B) and the elution was performed under a gradient mode at a flow rate of 1.0 mL/min. The detection wavelength was set at 300 nm. The HPLC-DAD method yielded a base line separation of the 11 components in 50% methanol extract of Radix Saposhnikoviae with no interfering peaks detected. The HPLC-DAD method was validated in terms of linearity, accuracy and precision (intra- and inter-day), limit of quantification (LOQ), recovery, and robustness. Specific determination of the 11 components was also accomplished by a triple quadrupole tandem mass spectrometer equipped with an electrospray ionization (ESI) source. This HPLC-MS/MS method was also validated by determining the linearity, limit of quantification, accuracy, and precision. Quantification of the 11 components in 51 commercial Radix Saposhnikoviae samples was successfully performed using the developed HPLC-DAD method. The identity, batch-to-batch consistency, and authenticity of Radix Saposhnikoviae were successfully monitored by the proposed HPLC-DAD and HPLC-MS/MS methods.
Product Description
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 2 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 10 mm, pk of 1 ea
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 300 μm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 3 mm, pk of
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 500 μm
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 3 mm, pk of
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea