Merck
  • Quantitative analysis of eletriptan in human plasma by HPLC-MS/MS and its application to pharmacokinetic study.
Quantitative analysis of eletriptan in human plasma by HPLC-MS/MS and its application to pharmacokinetic study.
Analytical and bioanalytical chemistry (2011-09-06)
Venkata Suresh Ponnuru, B R Challa, Ramarao Nadendla
ABSTRACT
Authors developed a simple, sensitive, selective, rapid, rugged, and reproducible liquid chromatography-tandem mass spectrometry method for the quantification of eletriptan (EP) in human plasma using naratriptan (NP) as an internal standard (IS). Chromatographic separation was performed on Ascentis Express C18, 50 × 4.6 mm, 2.7 μm column. Mobile phase was composed of 0.1% formic acid: methanol (40:60 v/v), with 0.5 mL/min flow rate. Drug and IS were extracted by liquid-liquid extraction. EP and NP were detected with proton adducts at m/z 383.2→84.3 and 336.2→97.8 in multiple reaction monitoring (MRM) positive mode, respectively. The method was validated with the correlation coefficients of (r(2)) ≥ 0.9963 over a linear concentration range of 0.5-250.0 ng/mL. This method demonstrated intra- and inter-day precision within 1.4-9.2% and 4.4-5.5% and accuracy within 96.8-103% and 98.5-99.8% for EP. This method is successfully applied in the bioequivalence study of 24 human volunteers.
Product Description
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 2 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 10 mm, pk of 1 ea
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 300 μm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 3 mm, pk of
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 500 μm
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 3 mm, pk of
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea