Merck
  • Molecular weight standards for calibration of gel filtration and sodium dodecyl sulfate-polyacrylamide gel electrophoresis: ferritin and apoferritin.
Molecular weight standards for calibration of gel filtration and sodium dodecyl sulfate-polyacrylamide gel electrophoresis: ferritin and apoferritin.
Analytical biochemistry (1987-11-01)
C de Haën
ABSTRACT
Ferritin and apoferritin are widely used for the calibration of gel filtration columns and sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis and are commercially offered for these purposes as part of molecular weight calibration kits. Many of the reported applications are severely in error as presented in leading references and application manuals. The manufacturers have based their recommendations on incorrect physicochemical parameters in the literature and incorrect or inadmissible assumptions about the protein subunit composition and architecture and have not taken into account the unusual resistance of these proteins to denaturation in SDS. Here the relevant physicochemical parameters of horse spleen apoferritin as reported in the literature are critically reevaluated and the best current estimates are identified as the following: weight average molecular weight of apoferritin, Mw = 481,200; molecular weight of subunits, major subunit, ML = 19,889; minor subunit, MH = 22,200; apparent specific volumes in 0.02 M acetate buffer, pH 5.5, and 0.1 M NaCl, phi = 0.721 ml g-1 and phi' = 0.743 ml g-1; partial specific volume at 20 degrees C, v = 0.738 ml g-1; viscosimetric molar volume, M[n] = 1.78 X 10(6) ml mol-1; Stokes radius, RSt = 67.1 A; viscosimetric radius, Rvis = 65.6 A; sedimentation coefficient S degrees 20, w = 16.6 S; translational diffusion coefficient, D20, w = 3.24 X 10(-7) cm2 s-1. Recommendations are provided for proper application of ferritin and apoferritin for calibration purposes in gel filtration and SDS-polyacrylamide gel electrophoresis.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Apoferritin from equine spleen, 0.2 μm filtered
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri