- N-myc downstream-regulated gene 1 promotes apoptosis in colorectal cancer via up-regulating death receptor 4.
N-myc downstream-regulated gene 1 promotes apoptosis in colorectal cancer via up-regulating death receptor 4.
Oncotarget (2017-11-16)
Xian Zhang, Bo Feng, Fan Zhu, Chaoran Yu, Jiaoyang Lu, Meng Pan, Zirui He, Xiongzhi Wangpu, Jing Sun, Xiao Yang
PMID29137287
ABSTRACT
The aim of this study was to evaluate the clinical significance of N-myc downstream-regulated gene 1 (NDRG1) in colorectal cancer (CRC) patients and to explore the mechanisms governing the role of NDRG1 in apoptosis of CRC cells. In the current study, we found that NDRG1 was a prognostic marker of CRC patients. Moreover, NDRG1 expression negatively correlated to tumor size and clinical TNM stage, suggesting that NDRG1 might act as a tumor suppressor by inhibiting proliferation or inducing apoptosis in CRC. Consistently, substantial apoptosis was observed