Merck
  • Highly selective and sensitive fluorogenic ferric probes based on aggregation-enhanced emission with -SiMe
Highly selective and sensitive fluorogenic ferric probes based on aggregation-enhanced emission with -SiMe
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-07-18)
Xuefeng Wang, Hua Wang, Qin Jiang, Yong-Ill Lee, Shengyu Feng, Hong-Guo Liu
ABSTRACT
In this study, thiophene was linked to polybenzene to generate novel fluorescent probes, namely 3,4-diphenyl-2,5-di(2-thienyl)phenyl-trimethylsilane (DPTB-TMS) with a -SiMe
Sigma-Aldrich
2-(Dimethylamino)pyridine, 97%
Sigma-Aldrich
Ethynyltrimethylsilane, 98%
Sigma-Aldrich
2-Thiopheneacetic acid, 98%