Merck
Δ
Biochimica et biophysica acta (2017-08-08)
Takuyu Hashiguchi, Katsuhisa Kurogi, Takehiko Shimohira, Takamasa Teramoto, Ming-Cheh Liu, Masahito Suiko, Yoichi Sakakibara
ABSTRACT
Cytosolic sulfotransferase (SULT)-mediated sulfation is generally known to involve the transfer of a sulfonate group from the active sulfate, 3'-phosphoadenosine 5'-phosphosulfate (PAPS), to a hydroxyl group or an amino group of a substrate compound. We report here that human SULT2A1, in addition to being able to sulfate dehydroepiandrosterone (DHEA) and other hydroxysteroids, could also catalyze the sulfation of Δ
Product Description
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)