- Does the fear avoidance model explain persistent symptoms after traumatic brain injury?
Does the fear avoidance model explain persistent symptoms after traumatic brain injury?
A minority of patients with mild traumatic brain injury (mTBI) experience a persistent symptom complex also known as post-concussion syndrome. Explanations for this syndrome are still lacking. To investigate if the fear avoidance model, including catastrophizing thoughts and fear avoidance behaviour, poses a possible biopsychosocial explanation for lingering symptoms and delay in recovery after traumatic brain injury (TBI) with special focus on mTBI. Cross-sectional study. 48 patients with TBI, of which 31 patients with mTBI, had persistent symptoms (mean time since injury 48.2 months); 92% of the entire sample fulfilled the criteria for post-concussion syndrome. catastrophizing, fear-avoidance, depression and post-concussion symptoms. High levels of catastrophizing were found in 10% and high levels of fear avoidance behaviour were found in 35%. Catastrophizing, fear avoidance behaviour, depressive symptoms and post-concussion symptoms correlated significantly with each other (p < 0.05). The fear-avoidance model proposes a possible explanation for persistent symptoms. Validation and normative data are needed for suitable measures of catastrophizing and fear avoidance of post-concussion symptoms after TBI. Longitudinal prospective cohort studies are needed to establish its causal and explanatory nature.