Merck
  • Targeted RP9 ablation and mutagenesis in mouse photoreceptor cells by CRISPR-Cas9.
Targeted RP9 ablation and mutagenesis in mouse photoreceptor cells by CRISPR-Cas9.
Scientific reports (2017-02-22)
Ji-Neng Lv, Gao-Hui Zhou, Xuejiao Chen, Hui Chen, Kun-Chao Wu, Lue Xiang, Xin-Lan Lei, Xiao Zhang, Rong-Han Wu, Zi-Bing Jin
ABSTRACT
Precursor messenger RNA (Pre-mRNA) splicing is an essential biological process in eukaryotic cells. Genetic mutations in many spliceosome genes confer human eye diseases. Mutations in the pre-mRNA splicing factor, RP9 (also known as PAP1), predispose autosomal dominant retinitis pigmentosa (adRP) with an early onset and severe vision loss. However, underlying molecular mechanisms of the RP9 mutation causing photoreceptor degeneration remains fully unknown. Here, we utilize the CRISPR/Cas9 system to generate both the Rp9 gene knockout (KO) and point mutation knock in (KI) (Rp9, c.A386T, P.H129L) which is analogous to the reported one in the retinitis pigmentosa patients (RP9, c.A410T, P.H137L) in 661 W retinal photoreceptor cells in vitro. We found that proliferation and migration were significantly decreased in the mutated cells. Gene expression profiling by RNA-Seq demonstrated that RP associated genes, Fscn2 and Bbs2, were down-regulated in the mutated cells. Furthermore, pre-mRNA splicing of the Fscn2 gene was markedly affected. Our findings reveal a functional relationship between the ubiquitously expressing RP9 and the disease-specific gene, thereafter provide a new insight of disease mechanism in RP9-related retinitis pigmentosa.
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Cone Arrestin Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit