Merck
  • 5-methylcytosine promotes mRNA export - NSUN2 as the methyltransferase and ALYREF as an m
5-methylcytosine promotes mRNA export - NSUN2 as the methyltransferase and ALYREF as an m
Cell research (2017-04-19)
Xin Yang, Ying Yang, Bao-Fa Sun, Yu-Sheng Chen, Jia-Wei Xu, Wei-Yi Lai, Ang Li, Xing Wang, Devi Prasad Bhattarai, Wen Xiao, Hui-Ying Sun, Qin Zhu, Hai-Li Ma, Samir Adhikari, Min Sun, Ya-Juan Hao, Bing Zhang, Chun-Min Huang, Niu Huang, Gui-Bin Jiang, Yong-Liang Zhao, Hai-Lin Wang, Ying-Pu Sun, Yun-Gui Yang
ABSTRACT
5-methylcytosine (m
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution