Merck
HPLC Analysis of Rapamycin and 32-Desmethoxyrapamycin on Discovery® HS C18
HPLC Analysis of Rapamycin and 32-Desmethoxyrapamycin on Discovery® HS C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery HS C18, 15 cm × 4.6 mm ID, 3μm particles (568522-U)
mobile phase
[A] 10 mM ammonium acetate, pH unadjusted: [B] acetonitrile (25:75, v/v, A:B)
flow rate
1.5 mL/min
column temp.
60 °C
detector
UV, 268 nm
injection
10 μg/mL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany