Produktname
Natrium-Tetrakis(4-fluorphenyl)borat Dihydrat ≥97.0% (NT)
Produktnummer
72016
Produktmarke
aldrich
CAS-Nummer
207683-22-5
Molekulargewicht
450.21
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| TITRATION (NT) HCLO4 0.1M | 97.0 - 103.0 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |