Produktname
Chlorophyll b analytical standard
Produktnummer
00538
Produktmarke
sial
CAS-Nummer
519-62-0
Molekulargewicht
907.47
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | DARK GREEN TO VERY DARK GREEN AND DARK BLUE TO VERY DARK BLUE |
| APPEARANCE (FORM) | Powder |
| PURITY (HPLC AREA %) | ≥ 95.0 % |
| TEST BY UV SPECTROSCOPY | CORRESPONDS TO REQUIREMENTS |
| EXPIRY PERIOD | 36 MONTHS |