Produktname
Butylbenzol analytical standard
Produktnummer
19600
Produktmarke
sial
CAS-Nummer
104-51-8
Molekulargewicht
134.22
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| PURITY (GC AREA %) | ≥ 99.8 % |
| REFRACTIVE INDEX N20/D | 1.489 - 1.491 |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |