Produktname
Imazalil PESTANAL®, analytical standard
Produktnummer
32007
Produktmarke
sial
CAS-Nummer
35554-44-0
Molekulargewicht
297.18
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| MELTING POINT | 50 - 55 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |