Produktname
Dimethylvinphos PESTANAL®, analytical standard
Produktnummer
32063
Produktmarke
sial
CAS-Nummer
2274-67-1
Molekulargewicht
331.52
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| MELTING POINT | 61 - 69 C |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |