Produktname
Buparvaquon VETRANAL®, analytical standard
Produktnummer
32154
Produktmarke
sial
CAS-Nummer
88426-33-9
Molekulargewicht
326.43
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Faint Yellow to Dark Yellow to Dark Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 98.0 % |
| MELTING POINT | 180 - 185 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |