Produktname
Sethoxydim PESTANAL®, analytical standard
Produktnummer
36795
Produktmarke
sial
CAS-Nummer
74051-80-2
Molekulargewicht
327.48
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Faint Yellow to Very Dark Yellow to Very Dark Brown-Yellow |
| APPEARANCE (FORM) | Liquid or Viscous Liquid |
| PURITY (HPLC AREA %) | ≥ 85.0 % |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |