Produktname
Phenylessigsäure analytical standard
Produktnummer
38166
Produktmarke
sial
CAS-Nummer
103-82-2
Molekulargewicht
136.15
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 99.0 % |
| CARBON CONTENT | 70.57 % (theory) |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 48 MONTHS |