Produktname
Tigloyl-L-carnitin analytical standard
Produktnummer
39588
Produktmarke
sial
CAS-Nummer
64681-36-3
Molekulargewicht
243.30
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 94.0 % |
| SPECIFIC ROTATION (20/D) | -20.0 - -16.0 Degrees |
| CONCENTRATION | c=0.1 in water |
| WATER | ≤ 10 % (calculated from elemental analysis) |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| CARBON 13 NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |