Produktname
Cycluron PESTANAL®, analytical standard
Produktnummer
45409
Produktmarke
sial
CAS-Nummer
2163-69-1
Molekulargewicht
198.31
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow to Faint Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| MELTING POINT | 135 - 140 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |