Produktname
Corticosteron VETRANAL®, analytical standard
Produktnummer
46148
Produktmarke
sial
CAS-Nummer
50-22-6
Molekulargewicht
346.46
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow to Faint Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| SPECIFIC ROTATION (20/D) | 219.0 - 226.0 Degrees |
| CONCENTRATION | C=1 IN ETHANOL |
| MELTING POINT | 180 - 189 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |