Produktname
Natriumtetraborat BioUltra, anhydrous, ≥99%
Produktnummer
71997
Produktmarke
sial
CAS-Nummer
1330-43-4
Molekulargewicht
201.22
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals |
| TITRATION (T) NAOH 1M | 98.5 - 101.4 % |
| LOSS ON DRYING | loss on drying ≤ 1 % 20C HV |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| ALUMINIUM (ICP) | ≤ 5 mg/kg |
| BARIUM (ICP) | ≤ 5 mg/kg |
| BISMUTH (ICP) | ≤ 5 mg/kg |
| CALCIUM (ICP) | ≤ 25 mg/kg |
| CADMIUM (ICP) | ≤ 5 mg/kg |
| COBALT (ICP) | ≤ 5 mg/kg |
| CHROMIUM (ICP) | ≤ 5 mg/kg |
| COPPER (ICP) | ≤ 5 mg/kg |
| IRON (ICP) | ≤ 5 mg/kg |
| POTASSIUM (ICP) | ≤ 50 mg/kg |
| LITHIUM (ICP) | ≤ 5 mg/kg |
| MAGNESIUM (ICP) | ≤ 5 mg/kg |
| MANGANESE (ICP) | ≤ 5 mg/kg |
| MOLYBDENUM (ICP) | ≤ 5 mg/kg |
| NICKEL (ICP) | ≤ 5 mg/kg |
| LEAD (ICP) | ≤ 5 mg/kg |
| STRONTIUM (ICP) | ≤ 5 mg/kg |
| ZINC (ICP) | ≤ 5 mg/kg |
| ARSENIC TRACES (MHS-AAS) | ≤ 0.5 mg/kg |
| TOTAL SULFUR AS SO4 (ICP) | ≤ 50 mg/kg |
| CHLORIDE (CL) | ≤ 10 mg/kg |
| PHOSPHATE (PO4) | ≤ 20 mg/kg |
| SOLUBILITY (METHOD) | 0.1 M in H2O |
| APPEARANCE (SOLUTION) | CLEAR COLORLESS |
| PH (SOLUTION) | 9.0 - 10.5 |
| RESIDUE (FILTER TEST) | NO RESIDUE |
| UV - ABS. AT 260 NM | ≤ 0.020 |
| UV - ABS. AT 280 NM | ≤ 0.015 |
| RECOMMENDED RETEST PERIOD | 72 MONTHS |