Produktname
2-Methyltetrahydrofuran analytical standard
Produktnummer
85488
Produktmarke
sial
CAS-Nummer
96-47-9
Molekulargewicht
86.13
Test | Spezifikation |
|---|---|
| STABILIZER | STABILIZED WITH ~0.025 % 2,6-DI-TERT-BUTYL-4-METHYLPHENOL |
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| PURITY (GC AREA %) | ≥ 99.5 % |
| REFRACTIVE INDEX N20/D | 1.405 - 1.407 |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |