Produktname
Kaliumsorbat purum p.a., ≥99.0% (NT)
Produktnummer
85520
Produktmarke
sial
CAS-Nummer
24634-61-5
Molekulargewicht
150.22
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow |
| APPEARANCE (FORM) | Beads |
| TITRATION (NT) HCLO4 0.1M | ≥ 99.0 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 1G IN 10ML WATER |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| CALCIUM (ICP) | ≤ 50 mg/kg |
| CADMIUM (ICP) | ≤ 50 mg/kg |
| COBALT (ICP) | ≤ 50 mg/kg |
| COPPER (ICP) | ≤ 50 mg/kg |
| IRON (ICP) | ≤ 50 mg/kg |
| SODIUM (ICP) | ≤ 3000 mg/kg |
| NICKEL (ICP) | ≤ 50 mg/kg |
| LEAD (ICP) | ≤ 50 mg/kg |
| ZINC (ICP) | ≤ 50 mg/kg |
| CHLORIDE (CL) | ≤ 50 mg/kg |