Produktname
Hydroxyethylcellulose
Produktnummer
09368
Produktmarke
sigma
CAS-Nummer
9004-62-0
Molekulargewicht
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Faint Beige |
| APPEARANCE (FORM) | Powder |
| LOSS ON DRYING | ≤ 7.0 % |
| RESIDUE ON IGNITION | ≤ 4.0 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| VISCOSITY (ROTATION) | 110 - 220 mPas |
| VISCOSITY (CONDITIONS) | 1% IN H2O AT 20 C, DIN 45, 100 S-1 |