Produktname
Ammoniumchlorid BioUltra, ≥99.5% (AT)
Produktnummer
09718
Produktmarke
sigma
CAS-Nummer
12125-02-9
Molekulargewicht
53.49
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals |
| ARGENTOMETR. TITRATION | 99.5 - 101.0 % |
| METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
| ALUMINIUM (ICP) | ≤ 5 mg/kg |
| BARIUM (ICP) | ≤ 5 mg/kg |
| BISMUTH (ICP) | ≤ 5 mg/kg |
| CALCIUM (ICP) | ≤ 10 mg/kg |
| CADMIUM (ICP) | ≤ 5 mg/kg |
| COBALT (ICP) | ≤ 5 mg/kg |
| CHROMIUM (ICP) | ≤ 5 mg/kg |
| COPPER (ICP) | ≤ 2 mg/kg |
| IRON (ICP) | ≤ 2 mg/kg |
| POTASSIUM (ICP) | ≤ 50 mg/kg |
| LITHIUM (ICP) | ≤ 5 mg/kg |
| MAGNESIUM (ICP) | ≤ 5 mg/kg |
| MANGANESE (ICP) | ≤ 5 mg/kg |
| MOLYBDENUM (ICP) | ≤ 5 mg/kg |
| SODIUM (ICP) | ≤ 50 mg/kg |
| NICKEL (ICP) | ≤ 1 mg/kg |
| LEAD (ICP) | ≤ 1 mg/kg |
| STRONTIUM (ICP) | ≤ 5 mg/kg |
| ZINC (ICP) | ≤ 2 mg/kg |
| ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
| NITRATE (NO3) | ≤ 5 mg/kg |
| PHOSPHATE (PO4) | ≤ 2 mg/kg |
| SULFATE (SO4) | ≤ 20 mg/kg |
| EXTRANEOUS ACTIVITIES | DNAses, RNAses, Proteases, Phosphatases Not Detectable |
| SOLUBILITY (METHOD) | water 1M |
| APPEARANCE (SOLUTION) | CLEAR COLORLESS |
| PH (SOLUTION) | 4.5 - 5.5 |
| RESIDUE (FILTER TEST) | NO RESIDUE |
| UV - ABS. AT 260 NM | ≤ 0.021 |
| UV - ABS. AT 280 NM | ≤ 0.019 |
| RECOMMENDED RETEST PERIOD | 72 MONTHS |